missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPSTI1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11627-1-AP-150UL
This item is not returnable.
View return policy
Description
EPSTI1 was up-regulated in breast carcinomas. The exact function of EPSTI1 is not known.Specifications
EPSTI1 | |
Polyclonal | |
Unconjugated | |
EPSTI1 | |
BRESI1, EPSTI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108670, 94240 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8VDI1, Q96J88 | |
EPSTI1 | |
EPSTI1 Fusion Protein Ag2157 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction