Learn More
Eppendorf™ Thermomixer™ R Accessory, Thermoblock Dry Blocks
Product Code. 15108374 Shop All Eppendorf Products
8 x 5.0 mL, Eppendorf Tube
Click to view available options
Holds:
8 x 5.0 mL
For Use With (Equipment):
Eppendorf Tube
Ensure precise fit for common lab tubes
These exchangeable thermoblocks for the Thermomixer R and ThermoStat provide excellent flexibility. The range of potential applications is significantly expanded due to the support for slides and microarrays, in addition to a broad range of tube sizes.
  • Shielded thermoblocks are contact-safe
  • Efficient exchange simplifies operation
  • Tubes are visible during operation and easily accessible
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Eppendorf Tube
Holds 8 x 5.0 mL
Type Block
For Use With (Application) For 8 x Eppendorf Tube 5.0 mL
Includes Not Applicable
Product Content Correction
Product Title
Eppendorf™ Thermomixer™ R Accessory, Thermoblock Dry Blocks >