Learn More
Eppendorf™ Eppendorf Quality™ epT.I.P.S.™ Set
Eppendorf™ Eppendorf Quality™ epT.I.P.S.™ Set are the perfect combination that results in reduced-force tip attachment and ejection with complete sealing.
Description
- Contamination-free transfer of trays to stable working box
- System optimized for use with multichannel pipettes
- Color-coded trays for simple identification of tips and pipette
- Tips can be attached to pipette from refill trays
- The trays and boxes can be autoclaved at 121°C for subsequent use