Learn More
Invitrogen™ Eotaxin-1 (CCL11) Mouse ELISA Kit
Sandwich ELISA Kit
Brand:  Invitrogen™ EMCCL11X5
 View more versions of this product
Product Code. 17115689
  • 2635.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Mouse Eotaxin-1/CCL11 ELISA quantitates Ms Eotaxin-1 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Eotaxin-1. Principle of the method The Mouse Eotaxin-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
AHC6054CU recognizes natural and recombinant eotaxin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P48298
1 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
20292
<12%
Standard, Pre-coated 96 well plates, Assay Diluent Concentrate, Stop Solution, Adhesive Plate Covers, SAV-HRP, Chromogen, Wash Buffer, Biotinylated Detection Antibody
5 x 96 Tests
Inflammation
2°C to 8°C
Mouse
4 hr. 45 min.
1.02 pg/mL to 100 pg/mL
1 pg/mL to 100 pg/mL
Biotin
Chemokine & Receptors
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF)
Mouse
Colorimetric Microplate Reader
Scya11,eotaxin
<10%
HRP
RUO
Plasma, 10 μ;L; Serum, 10 μ;L; Supernatant, 100 μ;L
Scya11, eotaxin
1 hr. 20 min.