Learn More
Cytiva Envirochek™ Supor™ Sampling Capsule
Envirochek™ capsules are approved for concentration and recovery of Cryptosporidium oocysts and Giardia cysts in water.
Brand: Cytiva 12110
This item is not returnable.
View return policy
Description
- Approved for concentration and recovery of Cryptosporidium oocysts and Giardia cysts in water according to EPA method 1623.1, with a faster, safer, and simpler protocol than other methods including processing of up to 8 samples at once using our shaker
Specifications
Urethane Adhesive | |
1 μm | |
1300 cm2 | |
1/Ea. |
Hydrophilic PES | |
Non-sterile | |
Polycarbonate | |
Straight Hose Barb |