Learn More
Lonza™ Clonetics™ Endothelial Cell Growth Medium BulletKit-2-MV
Used to culture microvascular endothelial cells. Lonza™ Clonetics™ Endothelial Cell Growth Medium BulletKit-2-MV is an optimized media for these primary cells.
Brand:  Lonza™ CC-3202
Product Code. 11605460
  • 500.00 € / Each
Explore more special offers
Description
Description
  • EGM™-2MV
  • Microvascular Endothelial Cell Growth Medium-2 BulletKit™
Order Info
dimProductType
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No BBE (bovine brain extract), hEGF, hydrocortisone, GA-1000 (Gentamicin, Amphotericin-B), FBS (fetal bovine serum) 25 ml, VEGF, hFGF-B, R3, -IGF-1, ascorbic acid
Growth Medium
1 Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • EGM-2 MV Endothel Med BulletKit
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P370+P378-In case of fire: Use to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, gentamycinsulphate