missing translation for 'onlineSavingsMsg'
Learn More

Endoglin/CD105 Mouse anti-Human, Unconjugated, Clone: 2D5E8, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen Endoglin/CD105
Clone 2D5E8
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16889485
View Documents
Proteintech
67075-1-IG-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16879485
View Documents
Proteintech
67075-1-IG-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Endoglin/CD105 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Endoglin/CD105. This antibody recognizes human antigen. The Endoglin/CD105 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.

Specifications

Specifications

Endoglin/CD105
1000 μg/mL
Monoclonal
Mouse
2022
Primary
2D5E8
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA
Unconjugated
Human
IgG2b
Videos
SDS
Documents

Documents

Product Certifications