Learn More
Merck SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holders
Mini and midi blot holders for SNAP i.d. 2.0 Protein Detection Systems
433.00 € - 525.00 €
Product Code | Brand | Length (English) Gel | Length (Metric) Gel | Width (English) Gel | Width (Metric) Gel | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) Gel | Length (Metric) Gel | Width (English) Gel | Width (Metric) Gel | Price | Quantity & Availability | ||
13288309
|
Merck
SNAP2BHMN0100 |
3.3 in. | 8.4 cm | 2.95 in. | 7.5 cm |
433.00 €
Pack of 100 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
13268309
|
Merck
SNAP2BHMD0100 |
5.31 in. | 13.5 cm | 3.34 in. | 8.5 cm |
525.00 €
Pack of 100 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
Description
For use with EMD Millipore SNAP i.d. 2.0 Protein Detection Systems (Cat. No. SNAP2MM)
Specifications
Blot Holder |