missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ Multiple Punch Kits
Includes punch with punch carrier plate and punch distributor
7250.00 €
Specifications
For Use With (Application) | Radionucleotide counting by transfer of individual filters to tubes |
---|---|
Quantity | 1/Pk. |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11556365
|
Merck
MAPK8960A |
Multiple punch, 2 carrier racks for 7mL vials |
7250.00 €
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
11566365
|
Merck
MAPK8960B |
Multiple punch, 2 carrier racks for 4mL vials |
7250.00 €
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
11722594
|
Merck
MAPK8960C |
Multiple punch, 1 carrier rack for 12 x 75mm tubes |
7250.00 €
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
Description
Specifications
Radionucleotide counting by transfer of individual filters to tubes | |
1/Pk. |