Learn More
Merck Millipore M Air T™ Agar Cassettes
Ready-to-use cassettes, prefilled with gamma-sterilized medium
Description
Description
  • Fast and reliable results with maximum recovery
  • Large agar amount and unique micro-perforated sieve allow air sampling of up to 1 cubic meter
  • Unit-to-unit consistency of agar surface shape
  • D
  • Reproducible results
Compliance
USP