missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Embelin, Thermo Scientific Chemicals
94.87 € - 308.00 €
Chemical Identifiers
CAS | 550-24-3 |
---|---|
Molecular Formula | C17H26O4 |
Molecular Weight (g/mol) | 294.39 |
MDL Number | MFCD00016369 |
InChI Key | IRSFLDGTOHBADP-UHFFFAOYSA-N |
Synonym | embelin, embelic acid, emberine, 2,5-dihydroxy-3-undecyl-1,4-benzoquinone, 2,5-dihydroxy-3-undecyl-p-benzoquinone, 2,5-dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione, unii-shc6u8f5er, embelin, embelia ribes, xiap inhibitor, embelin, shc6u8f5er |
PubChem CID | 3218 |
ChEBI | CHEBI:4778 |
IUPAC Name | 2,5-dihydroxy-3-undecylcyclohexa-2,5-diene-1,4-dione |
SMILES | CCCCCCCCCCCC1=C(O)C(=O)C=C(O)C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15496179
|
Thermo Scientific Alfa Aesar
J64074.MA |
10 mg |
94.87 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15406189
|
Thermo Scientific Alfa Aesar
J64074.MF |
50 mg |
308.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
An inhibitor of XIAP (X-linked inhibitor-of-apoptosis protein) that contains antitumor and anti-inflammatory activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- An inhibitor of XIAP (X-linked inhibitor-of-apoptosis protein) that contains antitumor and anti-inflammatory activity
Chemical Identifiers
550-24-3 | |
294.39 | |
IRSFLDGTOHBADP-UHFFFAOYSA-N | |
3218 | |
2,5-dihydroxy-3-undecylcyclohexa-2,5-diene-1,4-dione |
C17H26O4 | |
MFCD00016369 | |
embelin, embelic acid, emberine, 2,5-dihydroxy-3-undecyl-1,4-benzoquinone, 2,5-dihydroxy-3-undecyl-p-benzoquinone, 2,5-dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione, unii-shc6u8f5er, embelin, embelia ribes, xiap inhibitor, embelin, shc6u8f5er | |
CHEBI:4778 | |
CCCCCCCCCCCC1=C(O)C(=O)C=C(O)C1=O |
Specifications
550-24-3 | |
C17H26O4 | |
Light sensitive | |
embelin, embelic acid, emberine, 2,5-dihydroxy-3-undecyl-1,4-benzoquinone, 2,5-dihydroxy-3-undecyl-p-benzoquinone, 2,5-dihydroxy-3-undecyl-2,5-cyclohexadiene-1,4-dione, unii-shc6u8f5er, embelin, embelia ribes, xiap inhibitor, embelin, shc6u8f5er | |
CCCCCCCCCCCC1=C(O)C(=O)C=C(O)C1=O | |
294.39 | |
CHEBI:4778 | |
Solid |
Orange | |
MFCD00016369 | |
14,3555 | |
IRSFLDGTOHBADP-UHFFFAOYSA-N | |
2,5-dihydroxy-3-undecylcyclohexa-2,5-diene-1,4-dione | |
3218 | |
294.4 | |
Embelin |
Safety and Handling
missing translation for 'einecsNumber' : 208-979-8
missing translation for 'rtecsNumber' : DK4230000
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only