missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrolyte For Cadmium ISE

Brand:  Mettler Toledo™ 51107891

Product Code. 11957721

  • 49.75 € / 20mL
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Electrolyte for Cadmium ISE, 20mL
Electrolyte
20 mL
20 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers