Learn More
EDTA Disodium Salt Dihydrate, 99-101.0%, MB Grade, Ultrapure, Thermo Scientific Chemicals
Chelating agent used in many enzyme buffers and as enzyme inactivator at high concentrations
43.80 € - 126.00 €
Chemical Identifiers
CAS | 6381-92-6 |
---|---|
Molecular Formula | C10H18N2Na2O10 |
Molecular Weight (g/mol) | 372.238 |
MDL Number | MFCD00150037 |
InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
Synonym | edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs |
PubChem CID | 44120005 |
IUPAC Name | disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
SMILES | C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15865138
|
Thermo Scientific
J15701.22 |
100 g | Plastic bottle | ||||||
15720549
|
Thermo Scientific
J15701.36 |
500 g | Plastic bottle | ||||||
15815288
|
Thermo Scientific
J15701.A1 |
1 kg | Plastic bottle | ||||||
Description
Chelating agent used in many enzyme buffers and as an enzyme inactivator at high concentrations.
Stock Solution (0.5M):
- 186.1 gm EDTA
- H2O to 800 ml
- Titrate with approximately 20 gm of NaOH pellets to pH 8.0.
- Bring solution to 1 liter with water and sterilize by autoclaving.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6381-92-6 | |
372.238 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
44120005 | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+] |
C10H18N2Na2O10 | |
MFCD00150037 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
Specifications
248°C (decomposition) | |
Crystalline Powder | |
MFCD00150037 | |
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+] | |
372.238 | |
372.24 | |
Molecular Biology |
6381-92-6 | |
C10H18N2Na2O10 | |
3900609 | |
OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate | |
44120005 | |
99.0 to 101.0% | |
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate |
Safety and Handling
P260-P271-P304+P340-P312-P314-P501c
H332-H373
missing translation for 'einecsNumber' : 205-358-3
missing translation for 'rtecsNumber' : AH4375000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only