Learn More
Terumo™ Luer-Lock Tip Three-Part Syringe
Terumo™ Luer-Lock Tip Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
72.20 €
Specifications
Material | Polypropylene |
---|---|
Disposable | Single-use |
Connection Type | Luer Lock |
Graduated | Yes |
Sterility | Sterile |
Description
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Luer-Lock Three-Part Syringes feature a unique double beveled design for greater patient comfort.
- Silicone coated barrel for a smooth plunger action
- Clear barrel to allow quick recognition of air bubbles
- Easy to read markings
- Latex free
- Sterile and packed in individually wrapped tamper peel packs
- Syringes are available with luer lock, luer slip, catheter and eccentric tips
- Available in a wide range of sizes
Specifications
Polypropylene | |
Luer Lock | |
Sterile |
Single-use | |
Yes |
Safety and Handling
missing translation for 'shelfLife' : 5 Years