missing translation for 'onlineSavingsMsg'
Learn More

Eaton's reagent, Thermo Scientific Chemicals

112.56 € - 427.13 €

Specifications

Chemical Name or Material Eaton's reagent
Color Undesignated
CAS 39394-84-8, 75-75-2, 1314-56-3
Boiling Point 122.0°C (1.0 mmHg)
Molecular Formula O10P4
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10611473
View Documents Promotion Details
Thermo Scientific Acros
369805000
500 mL Glass bottle
112.56 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10423165
View Documents Promotion Details
Thermo Scientific Acros
369800025
2.5 L Glass bottle
427.13 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Eaton's reagent
39394-84-8, 75-75-2, 1314-56-3
O10P4
6.7 to 8.2 %
1.5000g/mL
283.89
Glass bottle
1.5
Liquid
Undesignated
122.0°C (1.0 mmHg)
MFCD00070545
05,535; 07,291; 08,400; 11,428
Phosphorus pentoxide, 7.5 wt.% solution in methanesulfonic acid
6.7 to 8.2%
1.4330 to 1.4360
>110°C
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. May be corrosive to metals. Harmful if swallowed. Harmful in contact with skin. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS Signal Word: Danger

Product Identifier
  • Eaton's reagent

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H290-May be corrosive to metals.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only