missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EasyStart™ PCR Mix-in-a-Tube
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Brand: Thermo Scientific™ 6024-11
This item is not returnable.
View return policy
Description
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Features:
- Delivers better specificity and yield than conventionally prepared mixtures
- Compatible with a wide range of enzymes
- Wax layer provides safe, easy storage of reagent mixture, prevents oxidation and evaporation, and eliminates wasted reagents
Ordering Information:
Use 0.2mL tubes for 0.2mL heating blocks and 0.5mL tubes for 0.5 or 0.6mL heating blocks.

Specifications
EasyStart Micro 100 | |
Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip | |
Tube |
0.2 mL | |
50 to 100 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ EasyStart™ PCR Mix-in-a-Tube > Micro 100; 0.2mL
Spot an opportunity for improvement?Share a Content Correction