missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester, 98%, Thermo Scientific™
72.71 € - 291.45 €
Chemical Identifiers
CAS | 185427-48-9 |
---|---|
Molecular Formula | C18H34B2O4 |
Molecular Weight (g/mol) | 336.09 |
MDL Number | MFCD03093898,MFCD03093898 |
InChI Key | SACOFNGKSPZAFQ-UHFFFAOYSA-N |
Synonym | e-1-hexene-1,2-diboronic acid bis pinacol ester, 1e-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-ene, z-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1-hexene, 4,4,5,5-tetramethyl-2-1z-2-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-z-1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane |
PubChem CID | 6026893 |
IUPAC Name | 4,4,5,5-tetramethyl-2-[(Z)-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hex-1-en-2-yl]-1,3,2-dioxaborolane |
SMILES | CCCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11315465
|
Thermo Scientific Alfa Aesar
L19650.03 |
1 g |
72.71 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11325465
|
Thermo Scientific Alfa Aesar
L19650.06 |
5 g |
291.45 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
185427-48-9 | |
336.09 | |
SACOFNGKSPZAFQ-UHFFFAOYSA-N | |
6026893 | |
CCCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1 |
C18H34B2O4 | |
MFCD03093898,MFCD03093898 | |
e-1-hexene-1,2-diboronic acid bis pinacol ester, 1e-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-ene, z-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1-hexene, 4,4,5,5-tetramethyl-2-1z-2-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-z-1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane | |
4,4,5,5-tetramethyl-2-[(Z)-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hex-1-en-2-yl]-1,3,2-dioxaborolane |
Specifications
185427-48-9 | |
102°C to 104°C (0.08 mmHg) | |
C18H34B2O4 | |
MFCD03093898,MFCD03093898 | |
e-1-hexene-1,2-diboronic acid bis pinacol ester, 1e-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-ene, z-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1-hexene, 4,4,5,5-tetramethyl-2-1z-2-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-z-1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-tetramethyl-1,3,2-dioxaborolan-2-yl hex-1-en-2-yl-1,3,2-dioxaborolane | |
CCCCC(=CB1OC(C)(C)C(C)(C)O1)B1OC(C)(C)C(C)(C)O1 | |
336.09 | |
336.09 | |
(E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester |
0.954 | |
>110°C (230°F) | |
1.456 | |
7768251 | |
SACOFNGKSPZAFQ-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-[(Z)-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hex-1-en-2-yl]-1,3,2-dioxaborolane | |
6026893 | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title