missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DZIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13779-1-AP-20UL
This item is not returnable.
View return policy
Description
May participate in spermatogenesis via its interaction with DAZ.Specifications
DZIP1 | |
Polyclonal | |
Unconjugated | |
DZIP1 | |
DAZ interacting protein 1, DAZ interacting protein 1/2, DZIP, DZIP1, DZIP2, DZIPt1, KIAA0996, RP11 23E3.3, Zinc finger protein DZIP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22873, 66573 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86YF9, Q8BMD2 | |
Dzip1 | |
DZIP1 Fusion Protein Ag4772 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |