Learn More
Dysprosium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals
For Use in Cermets Used As Nuclear Reaction Control Rods
53.90 € - 795.00 €
Chemical Identifiers
CAS | 1308-87-8 |
---|---|
Molecular Formula | Dy2O3 |
Molecular Weight (g/mol) | 373.00 |
MDL Number | MFCD00010986 |
InChI Key | GEZAXHSNIQTPMM-UHFFFAOYSA-N |
IUPAC Name | didysprosium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Dy+3].[Dy+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321079
|
Thermo Scientific Alfa Aesar
011319.09 |
10 g |
53.90 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301079
|
Thermo Scientific Alfa Aesar
011319.18 |
50 g |
204.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311079
|
Thermo Scientific Alfa Aesar
011319.30 |
250 g |
795.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In cermets used as nuclear reaction control rodsDysprosium trioxide is used with nickel in cermets, which are used as nuclear reactor control rods. It is also used in ceramics, glass, phosphors, lasers and dysprosium metal halide lamps. It is an important raw material to prepare dysprosium(III) salts by reacting with acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-87-8 | |
373.00 | |
GEZAXHSNIQTPMM-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Dy+3].[Dy+3] |
Dy2O3 | |
MFCD00010986 | |
didysprosium(3+) trioxidandiide |
Specifications
2340°C | |
White | |
99.9% (REO) | |
MFCD00010986 | |
GEZAXHSNIQTPMM-UHFFFAOYSA-N | |
didysprosium(3+) trioxidandiide | |
373 | |
White powder or lump | |
Dysprosium(III) oxide |
1308-87-8 | |
Powder | |
Dy2O3 | |
Insoluble in water. | |
[O--].[O--].[O--].[Dy+3].[Dy+3] | |
373.00 | |
Hygroscopic | |
7.81^2^7 |
Safety and Handling
missing translation for 'einecsNumber' : 215-164-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only