missing translation for 'onlineSavingsMsg'
Learn More
Dysprosium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™ Product Code.: 11394108

Dysprosium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™

Product Code. 11394108
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11394108

Brand: Thermo Scientific Alfa Aesar 041409.09

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13569-80-7
MDL Number MFCD00016071

Specifications

Boiling Point 2200°C
Melting Point 1360°C
Color White
Physical Form Pieces
Quantity 10 g
Solubility Information Soluble in strong mineral acids. Insoluble in water.
Formula Weight 219.5
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.5 g/mL
Assay (REO)
Chemical Name or Material Dysprosium(III) fluoride, Anhydrous
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dysprosium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.