Learn More
DUSP16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14237-1-AP-150UL
 View more versions of this product
Product Code. 16842784
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene encodes a mitogen-activated protein kinase phosphatase that is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. The encoded protein specifically regulates the c-Jun amino-terminal kinase and extracellular signal-regulated kinase pathways.
Specifications
Specifications
DUSP16
Polyclonal
Unconjugated
DUSP16
DUSP16, KIAA1700, MAP kinase phosphatase 7, MKP 7, MKP7
Rabbit
Antigen Affinity Chromatography
RUO
297682, 70686, 80824
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BY84
Dusp16
DUSP16 Fusion Protein Ag5479
150 μL
Primary
Human, Mouse, Rat