Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread
The threaded tube is used for further processing in glass components.
36.90 € - 113.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Borosilicate Glass (3.3) |
For Use With (Application) | Shoring in glass apparatuses |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
11742919
|
DWK Life Sciences
248360207 |
GL 14 |
39.70 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
11752919
|
DWK Life Sciences
248370105 |
GL 18 |
46.15 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
13167613
|
DWK Life Sciences
248350103 |
GL 45 |
36.90 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
11762919
|
DWK Life Sciences
248380209 |
GL 25 |
60.40 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
11772919
|
DWK Life Sciences
248390107 |
GL 32 |
113.00 €
Pack of 10 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||||
Description
This threaded tube has a screw thread and can be incorporated into glass components by a further processing step.
- With DIN thread
Specifications
Autoclavable | |
Borosilicate Glass (3.3) | |
Shoring in glass apparatuses |