Learn More
DWK Life Sciences DURAN™ Standard Socket, finished
For a rigid, self-supporting connection.
3.59 € - 394.00 €
Specifications
Type Finished
View More Specs
Products 17
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
12028184
View Documents
DWK Life Sciences
286550001
33 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15864828
View Documents
DWK Life Sciences
286561204
50 mm
126.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15814818
View Documents
DWK Life Sciences
286536602
22 mm
171.00 €
Pack of 50
In Stock
to see stock.
Add to basket
15854808
View Documents
DWK Life Sciences
286525408
17 mm
141.00 €
Pack of 50
In Stock
to see stock.
Add to basket
15814808
View Documents
DWK Life Sciences
286520007
15 mm
152.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15844828
View Documents
DWK Life Sciences
286559207
40 mm
92.90 €
Pack of 10
In Stock
to see stock.
Add to basket
15884798
View Documents
DWK Life Sciences
286515604
10 mm
43.30 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
10569381
View Documents
DWK Life Sciences
286561401
110 mm
111.00 €
Each
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15894798
View Documents
DWK Life Sciences
286517005
13 mm
163.00 €
Pack of 50
In Stock
to see stock.
Add to basket
12888041
View Documents
DWK Life Sciences
286543803
28 mm
229.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
10717691
View Documents
DWK Life Sciences
286561607
46 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15884828
View Documents
DWK Life Sciences
286563402
54 mm
243.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15894828
View Documents
DWK Life Sciences
286564407
58 mm
269.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15804838
View Documents
DWK Life Sciences
286565206
65 mm
302.00 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15814838
View Documents
DWK Life Sciences
286565806
75 mm
237.00 €
Pack of 5
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15824838
View Documents
DWK Life Sciences
286566202
90 mm
394.00 €
Pack of 5
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15874798
View Documents
DWK Life Sciences
286514608
8 mm
50.15 €
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
Description
Description
The conical ground joint is the most common joint in glass apparatus engineering. The DURAN™ sleeves, ground, comply with DIN 12249 Form E in their design and meet higher requirements for the surface roughness. This reduces the need for grinding grease and the risk of jamming. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • According to DIN 12249 Form E
  • Ground (finer grinding quality, less roughness depth)
  • The finer surface reduces the risk of jamming the cuts
  • Minimized grease requirement
  • Very low leakage rate
  • Tight closure due to standard ground joint
  • Glass articles for further processing
Specifications
Specifications
Finished