Learn More
DWK Life Sciences DURAN™ Standard Socket, double, diamond-ground
Designed for a rigid, self-supporting connection with lower requirements.
Brand:  DWK Life Sciences 286573309
 View more versions of this product
Product Code. 10131411
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
DURAN™ DG double sleeves are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ sleeves (not according to DIN)
  • Glass article with two sleeves enables material savings
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25/Pk.
Installation in glass apparatus and devices
34 mm
2 mm