Learn More
DWK Life Sciences DURAN™ Standard Cone, double, finished
For a rigid, self-supporting connection.
246.00 € - 487.00 €
Products 4
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15844598
View Documents
DWK Life Sciences
286172809
13 mm
304.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15854598
View Documents
DWK Life Sciences
286173205
17 mm
385.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15921541
View Documents
DWK Life Sciences
286174407
26 mm
246.00 €
Pack of 25
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15864598
View Documents
DWK Life Sciences
286174004
22 mm
487.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
Description
Description
The DURAN™ double cores, ground, are an old, less common type of design according to DIN 12249 Form D. Their primary purpose is to save material.
  • According to DIN 12249 Form D
  • Ground (finer grinding quality, less roughness depth)
  • Glass article with two cores enables material savings
  • Tight closure due to standard ground joint
  • Glass articles for further processing