missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
Click to view available options
Thread Size (Metric):
GL14
GL 18
GL 25
GL 32
GL 45
Description
Suitable for PBT open topped screw cap with central aperture Heat resistance: 130°C (steam) and 200°C (dry heat). Silicon is peroxide catalysed.
- Compatible with open topped GL screw caps with central aperture
- Compatible with GL bottles
- Peroxide catalysed silicone
Specifications
Specifications
For Use With (Application) | Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments |
Material | Silicone |
Quantity | 100/Pk. |
Thickness (Metric) | 2 mm |
Thread Size (Metric) | GL 32 |
Autoclavable | Autoclavable |
Diameter (Metric) | 29 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'