Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, SVL
To attach.
Brand:  DWK Life Sciences 286064408
140.20 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15844548
  • 162.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
The DURAN™ glass thread tube with SVL thread is a finer thread than the GL thread, which is also used for attaching to glass apparatus. The nominal size refers to the core diameter at the thread.
  • Glass thread tube with SVL thread is a finer thread compared to GL thread
  • The nominal size refers to the core diameter at the thread.
  • Safe and easy sealing and loosening of glass-glass components
  • Grease-free working possible
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
40 mm
2 mm
For attachment to glass apparatuses; Installation in glass apparatus and devices
SVL 42