Learn More
DWK Life Sciences DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated
Description
The DURAN™ premium GL 45 laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The premium bottles are ideal for high temperature applications: the thermal shock resistance of the glass bottle is certified for a 160°C differential, and is combined with a high temperature fluoropolymer (PFA) screw cap and pouring ring that are suitable for use up to 200°C. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature a double graduated scale for filling volume and a reversed scale for pouring. The marking field and Retrace Code for manufacturing lot traceability are printed in permanent white enamel.
- Additional graduation marks and an opposite scaling (accurate to ± 5 %) simplify the reading of the filling level as well as the quantity withdrawn
- Colourant free product
- Reliable sealing due to PTFE-coated cap liner
- TÜV tested thermal shock resistance of 160 K
- Complete with PFA pouring ring and cap
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
Specifications
Specifications
Capacity (Metric) | 250 mL |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping |
Includes | With premium screw cap and pouring ring |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
Closure Material | PFA/PFA, PTFE/Silicone Layer |
Diameter (Metric) | 70 mm |