missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Original Laboratory GL Bottle Screw Cap, Linerless Lip Seal, PP

Product Code. 11722322
Click to view available options
Thread Size (Metric):
GL 25
GL 32
GL 45
Color:
Blue
Gray
Green
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Screw cap with reliable plug seal.

The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.

  • Cap with integral lip seal provides effective sealing for short term storage
  • Pouring ring for drip-free pouring
  • The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
  • Fully compatible with all the GL 45 threads and therefore highly versatile
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Description Screw cap
For Use With (Application) Autoclaving media and reagents; Short term sample and media storage; color coding and identification
Material Polypropylene
Quantity 10/Pk.
Thread Size (Metric) GL 25
Autoclavable Autoclavable
Diameter (Metric) 33 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.