missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Original GL 45 Laboratory Bottle, with High Temperature Screw cap, Clear, with DIN 168-1 Thread, Clear, Graduated

Product Code. 15876871
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

DURAN™ Original GL 45 Laboratory Bottles complete with High Temperature Closures.

DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN Original GL 45 laboratory bottles are available complete with high temperature screw caps and pouring rings. The red polybutylene terephthalate (PBT) GL 45 threaded caps with PTFE faced silicone cap liner and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. All components are fully autoclavable at 121 or 134°C and suitable for dry heat sterilization up to 180°C. All components are fully compliant with current international food contact regulations such as (EU) No 10/2011.

  • Supplied complete with red polybutylene terephthalate (PBT) GL 45 threaded cap with PTFE faced silicone liner and ETFE pouring ring for drip free pouring.
  • Supplied complete with high temperature PBT screw caps and ETFE pouring rings.
  • The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel.
  • Permanent Retrace Code on each bottle for full manufacturing lot traceability.
  • Suitable for long-term storage.
  • All components are fully autoclavable at 121 or 134°C.
  • All components are suitable for dry heat sterilization up to 180°C.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
For Use With (Application) Suitable for dry heat sterilization; Autoclaving of liquid media; Storage of corrosive reagents; Sampling; Storage of media and reagents; Transport and shipping
Includes With high temperature resistant PBT screw cap and ETFE pouring ring
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Thread Size (Metric) GL 45
Autoclavable Autoclavable
Closure Material PBT with ETFE Pouring Ring
Diameter (Metric) 86 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'