Learn More
DWK Life Sciences DURAN™ Long Ground Cone with tip, finished
For ground joints in the high-vacuum range with extension.
Brand:  DWK Life Sciences 286164809
 View more versions of this product
Product Code. 15834588
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The DURAN™ long ground cores with outlet combine the advantages of long ground and outlet cores.
  • Ground (finer grinding quality, less roughness depth)
  • The larger ground surface is suitable for use in high-vacuum applications.
  • With drain
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
13 mm
1.5 mm
The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe; Installation in glass apparatus and devices
NS 14/35