missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Round Bottom Four-Neck Flask

Especially made for reactions and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Round Bottom Four-Neck Flasks are manufactured from durable DURAN™ glass.

Brand:  Lenz Laborglasinstrumente™ 03344458

Product Code. 10728461

  • 116.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN 12392
  • With conical side joints
  • 3 parallel side necks at 90° angle
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

DIN 12392
Borosilicate Glass, Duran Glass
1 Each
Round
500 mL
Four-neck
NS 29/32
105 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.