Learn More
DWK Life Sciences DURAN™ Expansion Adapter, socket/cone
Expansion from sleeve to core.
Brand:  DWK Life Sciences 286549206
 View more versions of this product
Product Code. 15814828
  • 836.00 € / Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Adapters are used to connect two components with different NS sizes. DURAN™ expansion pieces allow the connection of a smaller sleeve to a larger core.
  • Expansion pieces allow the connection of a smaller sleeve to a larger core and thus the use of differently sized ground joint components.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Connection of two components with different standard ground joint sizes
Socket NS 29/32, Cone NS 71/51