Learn More
DuraCal™ 4.01 Value pH Buffer, Hamilton™ Product Code.: 10214122
DuraCal™ 4.01 Value pH Buffer, Hamilton™
Product Code. 10214122
500 mL
Click to view available options
Quantity:
500 mL
Product Code. 10214122
Brand: Hamilton 238217
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 4.01 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.
  • 4.01 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards
Specifications
pH 4.01
Quantity 500 mL
Product Content Correction
Product Title
DuraCal™ 4.01 Value pH Buffer, Hamilton™ >