missing translation for 'onlineSavingsMsg'
Learn More

DuraCal™ 2.00 Value pH Buffer, Hamilton™

Product Code. 10747444
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 2.00 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.

  • 2.00 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards

Specifications

pH 2.00
Quantity 500 mL
compliance-icons
Product Identifier
  • pH Buffer 2.00
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.