Learn More
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug
Made from borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug is used in the laboratory to transfer fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.
Brand:  Lenz Laborglasinstrumente™ 04457004
 View more versions of this product
Product Code. 11876114
  • 117.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conical shape with NS socket and NS cone
  • With solid glass plug, with screwthread retaining device
  • With inscription label, without scale
  • Borosilicate glass
Specifications
Specifications
1000 mL
Borosilicate Glass
6.0mm
1