missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Promotion
Contains green buffer that includes density reagent and two tracking dyes for direct loading of PCR products on gels
96.25 € - 383.00 €
Specifications
For Use With (Application) | Standard PCR, RT-PCR, Genotyping, Cloning |
---|---|
Concentration | 2X |
Product Code | Brand | Quantity | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | No. of Reactions | Price | Quantity & Availability | |||||
15717526
|
Thermo Scientific™
K1082 |
20 x 1.25mL | 1000 | N/A | ||||||
15707526
|
Thermo Scientific™
K1081 |
4 x 1.25 mL | 200 | N/A | ||||||
Description
- DreamTaq DNA Pol,
- dNTPs,
- Green buffer, density reagent and dyes for direct loading,
- supplied with Water,
- Nuclease free
Specifications
Standard PCR, RT-PCR, Genotyping, Cloning | |
2X |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Promotion