missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24063-1-AP-150UL
This item is not returnable.
View return policy
Description
TRAIL-R1 (CD261, DR4) is a type I transmembrane protein, also called TRAIL receptor 1. The ligand for this DR4 death receptor has been identified and termed TRAIL, which is a member of the TNF family. DR4, as many other receptors (Fas, TNFR1, etc.), mediates apoptosis and NF kappaB activation in many cells and tissues. Apoptosis, a programmed cell death, is a operating process in normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by coupled of certain cytokines (TNF family - TNF, Fas ligand) and their death domain containing receptors (TNFR1, Fas receptor).Specifications
DR4 | |
Polyclonal | |
Unconjugated | |
TNFRSF10A | |
APO2, CD261, Death receptor 4, DR4, TNFRSF10A, TRAIL R1, TRAIL receptor 1, TRAILR 1, TRAILR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8797 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00220 | |
TNFRSF10A | |
DR4 Fusion Protein Ag20927 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction