Learn More
Sampling Systems™ Double-ended Spoon
These double-ended spoons are ideal for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary, allowing for flexibility in sampling both powders and liquids.
Brand: Sampling Systems™ A104-300
This item is not returnable.
View return policy
Description
- Made from stainless steel
- Ideal for sampling powders and liquids
- Various sizes available
Specifications
Spoon | |
Autoclavable | |
No | |
300 mm |
Stainless Steel | |
Reusable | |
Non-sterile |