Learn More
Sampling Systems™ Double Bagged - USP Class VI Pallet Knives
BIOPHARMA
Our Double Bagged Pallet Knives are made from high grade medical USP Class VI HDPE. They are available with a handy 200 mm blade. This premium range is double bagged. A box liner is also used giving a third layer of protection.
Brand:  Sampling Systems™ 9175H-200S
Product Code. 17504706
  • 255.00 € / Pack of 40
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The SteriWare™ Ultra Pallet Knives are commonly used in the manufacture of cosmetics, food and pharmaceuticals.
  • They are also popular for levelling off scoops etc.
  • The pallet knives all have an ergonomic handle and are designed to flex.
  • The pallet knives are designed to be used straight from the bag.
  • Made from food grade, FDA and EC compliant HDPE.
  • HDPE has good chemical resistance and can be easily recycled.
  • The pallet knives are sterilised by gamma irradiation.
  • USP CLASS VI - High quality medical grade polymer that has also been independently tested to USP 661.
  • Cleanroom Manufactured - The pallet knives are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
  • Each pallet knife is 100% inspected before packing.
Specifications
Specifications
HDPE
Sterile
Double Bagged
200 mm