Learn More
Miele Professional™ Dosing system
Dispenser module DOS K 85 for 5-10 litre canisters with level indicator for the automatic dispensing of liquid cleaning agents (PG 8583/93)
Brand:  Miele Professional™ 9961070
Product Code. 15418701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Dispenser monitoring – in accordance with DIN EN ISO 15883
  • For dispensing liquid cleaning agent or neutralising agent
  • For dispensing chemical disinfectant
  • 333 mm siphon for 5-10 l containers
  • Conversion kit for siphons for 10-30 l canisters available
Specifications
Specifications
Dispenser
5-10 litre canisters