missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dopamine hydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 243°C to 250°C |
Assay Percent Range | 99% |
Quantity | 100 g |
Beilstein | 3656720 |
Sensitivity | Hygroscopic; Light sensitive |
Merck Index | 14,3421 |
Solubility Information | Soluble in water,methanol and dimethyl sulfoxide. Insoluble in ether,petroleum ether,chloroform,benzene and toluene. |
Formula Weight | 189.64 |
Percent Purity | 99% |
Chemical Name or Material | Dopamine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'