Learn More
Dodecylamine, 98%, Thermo Scientific Chemicals
38.90 € - 126.00 €
Chemical Identifiers
CAS | 124-22-1 |
---|---|
Molecular Formula | C12H27N |
Molecular Weight (g/mol) | 185.36 |
MDL Number | MFCD00008154 |
InChI Key | JRBPAEWTRLWTQC-UHFFFAOYSA-N |
Synonym | dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 |
PubChem CID | 13583 |
IUPAC Name | dodecan-1-amine |
SMILES | CCCCCCCCCCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10122960
|
Thermo Scientific Acros
117661000 |
100 g | Glass bottle |
38.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10549430
|
Thermo Scientific Acros
117665000 |
500 g | Glass bottle |
76.60 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10469410
|
Thermo Scientific Acros
117660010 |
1 kg | Glass bottle |
126.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-22-1 | |
185.36 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
13583 | |
CCCCCCCCCCCCN |
C12H27N | |
MFCD00008154 | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
dodecan-1-amine |
Specifications
124-22-1 | |
White | |
247.0°C to 249.0°C | |
Authentic | |
C12H27N | |
MFCD00008154 | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
dodecan-1-amine | |
13583 | |
98% | |
Dodecylamine, 98% |
28.0°C to 32.0°C | |
0.8 | |
>110°C | |
97.5% min. (Alkyl chain distribution C12) (GC) | |
CH3(CH2)11NH2 | |
04, 200 | |
Solubility in water: insoluble. Other solubilities: soluble in chloroform, isopropylalcohol, ethanol,, ether | |
CCCCCCCCCCCCN | |
185.36 | |
185.35 | |
Crystalline Low Melting Solid |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquat
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-690-6
missing translation for 'rtecsNumber' : JR6475000
missing translation for 'tsca' : TSCA
- Dodecylamine
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only