missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17043-1-AP-20UL
This item is not returnable.
View return policy
Description
GTPase which can activate the MEK/ERK pathway and induce cell transformation when overexpressed. May act as a nuclear scaffold for MAPK1, probably by association with MAPK1 nuclear export signal leading to enhanced ERK1/ERK2 signaling.Specifications
DNAJC27 | |
Polyclonal | |
Unconjugated | |
DNAJC27 | |
DNAJC27, RabJS, RBJ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217378, 298859, 51277 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6IML7, Q8CFP6, Q9NZQ0 | |
DNAJC27 | |
DNAJC27 Fusion Protein Ag10586 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction