missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB6 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1A1D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66587-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the DNAJ protein family. DNAJ family members are characterized by a highly conserved amino acid stretch called the ′J-domain′ and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events, such as protein folding and oligomeric protein complex assembly. This family member may also play a role in polyglutamine aggregation in specific neurons. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described.Specifications
DNAJB6 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O54946, O75190, Q6AYU3 | |
Dnajb6 | |
DNAJB6 Fusion Protein Ag2258 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A1D2 | |
Unconjugated | |
DNAJB6 | |
DJ4, DnaJ, DNAJB6, Heat shock protein J2, HHDJ1, HSJ 2, HSJ2, MRJ, MSJ 1, MSJ1 | |
Mouse | |
Protein G | |
RUO | |
10049, 100525669, 23950, 362293 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction