Learn More
Thermo Scientific Chemicals DL-Serine, 99%
55.98 € - 207.03 €
Chemical Identifiers
CAS | 302-84-1 |
---|---|
Molecular Formula | C3H7NO3 |
Molecular Weight (g/mol) | 105.093 |
MDL Number | MFCD00064223 |
InChI Key | MTCFGRXMJLQNBG-UHFFFAOYSA-N |
Synonym | dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid |
PubChem CID | 617 |
ChEBI | CHEBI:17822 |
IUPAC Name | 2-amino-3-hydroxypropanoic acid |
SMILES | C(C(C(=O)O)N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482188
|
Thermo Scientific Chemicals
A15184.22 |
100 g |
55.98 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492188
|
Thermo Scientific Chemicals
A15184.36 |
500 g |
207.03 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
302-84-1 | |
105.093 | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
617 | |
2-amino-3-hydroxypropanoic acid |
C3H7NO3 | |
MFCD00064223 | |
dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acidShow More | |
CHEBI:17822 | |
C(C(C(=O)O)N)O |
Specifications
∼240°C (decomposition) | |
C3H7NO3 | |
1721402 | |
dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid | |
MTCFGRXMJLQNBG-UHFFFAOYSA-N | |
2-amino-3-hydroxypropanoic acid | |
617 | |
105.09 | |
DL-Serine |
302-84-1 | |
MFCD00064223 | |
14,8460 | |
Soluble in water. | |
C(C(C(=O)O)N)O | |
105.093 | |
CHEBI:17822 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 206-130-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.