Learn More
DL-Lactic acid, 90%, Thermo Scientific Chemicals
27.34 € - 345.00 €
Chemical Identifiers
CAS | 50-21-5, 7732-18-5 |
---|---|
Molecular Formula | C3H6O3 |
Molecular Weight (g/mol) | 90.08 |
MDL Number | MFCD00004520 |
InChI Key | JVTAAEKCZFNVCJ-UHFFFAOYNA-N |
Synonym | lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum |
PubChem CID | 612 |
ChEBI | CHEBI:78320 |
IUPAC Name | 2-hydroxypropanoic acid |
SMILES | CC(O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10736341
|
Thermo Scientific Acros
125060250 |
25 g | Glass bottle |
27.34 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10153460
|
Thermo Scientific Acros
125065000 |
500 g | Glass bottle |
42.11 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031120
|
Thermo Scientific Acros
125060010 |
1 kg | Glass bottle |
66.03 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10487510
|
Thermo Scientific Acros
125060025 |
2.5 kg | Glass bottle |
138.69 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10554074
|
Thermo Scientific Acros
125060100 |
10 kg | Plastic drum |
345.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
50-21-5, 7732-18-5 | |
90.08 | |
JVTAAEKCZFNVCJ-UHFFFAOYNA-N | |
612 | |
2-hydroxypropanoic acid |
C3H6O3 | |
MFCD00004520 | |
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum | |
CHEBI:78320 | |
CC(O)C(O)=O |
Specifications
50-21-5, 7732-18-5 | |
122.0°C (15.0 mmHg) | |
C3H6O3 | |
MFCD00004520 | |
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum | |
JVTAAEKCZFNVCJ-UHFFFAOYNA-N | |
2-hydroxypropanoic acid | |
612 | |
90.08 | |
1.2000g/mL | |
Authentic | |
1.4252 to 1.4272 | |
1.2 |
18.0°C | |
Colorless to Yellow | |
CH3CH(OH)CO2H | |
15, 5384 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, furfurol, glycerol, less soluble in ether, practically insoluble in chloroform, carbon, disulfide, petroleum ether | |
CC(O)C(O)=O | |
90.08 | |
CHEBI:78320 | |
90% | |
>110°C | |
Liquid | |
0.1% max. | |
DL-Lactic acid |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.