missing translation for 'onlineSavingsMsg'
Learn More

DL-Adrenaline, Thermo Scientific Chemicals

Product Code. 15415289
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A hormone and neurotransmitter

A hormone and neurotransmitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329-65-7
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00063027
InChI Key UCTWMZQNUQWSLP-UHFFFAOYSA-N
Synonym dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
PubChem CID 838
ChEBI CHEBI:33568
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNCC(C1=CC(=C(C=C1)O)O)O

Specifications

Melting Point 197°C (decomposition)
Color White
Linear Formula (HO)2C6H3CH(CH2NHCH3)OH
Quantity 25 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,3619
Formula Weight 183.2
Physical Form Powder
Chemical Name or Material DL-Adrenaline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.