missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-10-Camphorsulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
16.70 € - 205.02 €
Chemical Identifiers
CAS | 34850-66-3 |
---|---|
Molecular Formula | C10H15NaO4S |
Molecular Weight (g/mol) | 254.28 |
MDL Number | MFCD00135623 |
InChI Key | AWMAOFAHBPCBHJ-UHFFFAOYSA-M |
Synonym | sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate |
PubChem CID | 23686666 |
IUPAC Name | sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate |
SMILES | CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10119981
|
Thermo Scientific Acros
189120050 |
5 g | Glass bottle |
16.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10477560
|
Thermo Scientific Acros
189125000 |
500 g | Plastic bottle |
205.02 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34850-66-3 | |
254.28 | |
AWMAOFAHBPCBHJ-UHFFFAOYSA-M | |
23686666 | |
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+] |
C10H15NaO4S | |
MFCD00135623 | |
sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate | |
sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate |
Specifications
34850-66-3 | |
White | |
98% | |
MFCD00135623 | |
Solubility in water: soluble | |
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)[O-])C.[Na+] | |
254.28 | |
254.28 | |
Amorphous Powder |
286°C to 288°C | |
Authentic | |
C10H15NaO4S | |
sodium camphorsulfonate, sodium 7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonate, sodium +/--10-camphorsulfonate, d-camphor-10-sulfonic acid sodium salt, sodium camphorsulfonate anion, bicyclo 2.2.1 heptane-1-methanesulfonic acid, 7,7-dimethyl-2-oxo-, sodium salt, +--sodium camphorsulfonate, sodium 2-oxobornane-10-sulphonate, sodium di-10-camphorsulfonate | |
AWMAOFAHBPCBHJ-UHFFFAOYSA-M | |
sodium;(7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl)methanesulfonate | |
23686666 | |
98% | |
DL-10-Camphorsulfonic acid, sodium salt, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 252-250-7
missing translation for 'tsca' : TSCA
Product Identifier
- DL-10-Camphorsulfonic acid, sodium salt
RUO – Research Use Only