missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-1,4-Dithiothreitol, 98%, Pure

43.62 € - 1259.27 €

Chemical Identifiers

CAS 3483-12-3
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.25
MDL Number MFCD00004877
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10792782
View Documents Promotion Details
Thermo Scientific Chemicals
409190010
1 g Glass bottle
43.62 €
1g
Please to purchase this item. Need a web account? Register with us today!
10686841
View Documents Promotion Details
Thermo Scientific Chemicals
409190050
5 g Glass bottle
111.56 €
5g
Please to purchase this item. Need a web account? Register with us today!
10424615
View Documents Promotion Details
Thermo Scientific Chemicals
409190250
25 g Glass bottle
446.22 €
25g
Please to purchase this item. Need a web account? Register with us today!
10434615
View Documents Promotion Details
Thermo Scientific Chemicals
409191000
100 g Glass bottle
1259.27 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3483-12-3
154.25
VHJLVAABSRFDPM-IMJSIDKUSA-N
446094
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
C4H10O2S2
MFCD00004877
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
CHEBI:42170
C(C(C(CS)O)O)S

Specifications

3483-12-3
White
>110°C
97.5 to 102.5% (Iodimetry)
HSCH2CH(OH)CH(OH)CH2SH
01, III, 2360
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
VHJLVAABSRFDPM-IMJSIDKUSA-N
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
446094
154.25
Pure
DL-1, 4-Dithiothreitol
39.0°C to 44.0°C
125.0°C to 130.0°C (2.0 mmHg)
Authentic
C4H10O2S2
MFCD00004877
15, 3419
Solubility in water: freely soluble. Other solubilities: freely soluble in ethanol,acetone,ether,,chloroform and ethyl acetate
C(C(C(CS)O)O)S
154.25
CHEBI:42170
98%
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 222-468-7

Product Identifier
  • DL-1,4-Dithiothreitol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only